accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNNOO

CAS RN 4835-39-6

IUPAC Name: N-(4-nitrophenyl)-3-oxobutanamide
Synonyms: N-(4-nitrophenyl)-3-oxobutanamide
Molecular Weight (g/mol): 222.2
Molecular Formula: C10H10N2O4
InChi Key: KCXJQNDNGLRYBN-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results