accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOFF

CAS RN 4837-20-1

IUPAC Name: 4-(difluoromethoxy)benzoate
Synonyms: 4-(difluoromethoxy)benzoate
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H5F2O3
InChi Key: BSNNYLYELGBSBA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(OC(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results