accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOH3C

CAS RN 4841-84-3

IUPAC Name: 1,2-dimethyl cyclohex-4-ene-1,2-dicarboxylate
Synonyms: 1,2-dimethyl cyclohex-4-ene-1,2-dicarboxylate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: DVVAGRMJGUQHLI-UHFFFAOYNA-N
SMILES: COC(=O)C1CC=CCC1C(=O)OC
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results