accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4845-58-3

IUPAC Name: 6-nitro-2,3-dihydro-1,3-benzothiazole-2-thione
Synonyms: 6-nitro-3H-1,3-benzothiazole-2-thione
Molecular Weight (g/mol): 212.24
Molecular Formula: C7H4N2O2S2
InChi Key: QPOZGXKWWKLJDK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)