accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOO

CAS RN 4857-42-5

IUPAC Name: 3-methyl-1,2-oxazole-5-carboxylate
Synonyms: 3-methyl-1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 126.09
Molecular Formula: C5H4NO3
InChi Key: HXIYCKAAQPHZBM-UHFFFAOYSA-M
SMILES: CC1=NOC(=C1)C([O-])=O
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results