accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 485799-04-0

IUPAC Name: 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
Synonyms: 4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
Molecular Weight (g/mol): 290.17
Molecular Formula: C15H23BN2O3
InChi Key: ZGDLVKWIZHHWIR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results