accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH

CAS RN 486-84-0

IUPAC Name: 1-methyl-9H-pyrido[3,4-b]indole
Synonyms: harmane 2-methyl-β-carboline 3-methyl-4-carboline pyridobindole, l-methyl- l-methyl-pyridobindole L-methylpyridobindole 1-methylnorharman
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: PSFDQSOCUJVVGF-UHFFFAOYSA-N
SMILES: CC1=NC=CC2=C1NC1=C2C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results