CAS RN 4861-79-4
IUPAC Name:
magnesium(2+) methanolate methyl carbonate
Synonyms:
magnesium(2+) methoxide methyl carbonate
Molecular Weight (g/mol):
130.38
Molecular Formula:
C3H6MgO4
InChi Key:
PNVKCUSCADAAMP-UHFFFAOYSA-M
SMILES:
[Mg++].C[O-].COC([O-])=O
1
–
1
of
1
results
1
Sigma Aldrich Magnesium Methyl Carbonate Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.