accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4865-84-3

IUPAC Name: 2-(1,2-benzoxazol-3-yl)acetate
Synonyms: 2-(1,2-benzoxazol-3-yl)acetate
Molecular Weight (g/mol): 176.15
Molecular Formula: C9H6NO3
InChi Key: BVSIAYQIMUUCRW-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=NOC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results