accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHS

CAS RN 4869-59-4

IUPAC Name: 3-sulfanylbenzoic acid
Synonyms: 3-sulfanylbenzoic acid
Molecular Weight (g/mol): 154.18
Molecular Formula: C7H6O2S
InChi Key: RSFDFESMVAIVKO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(S)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results