accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3OO

CAS RN 487-06-9

IUPAC Name: 5,7-dimethoxy-2H-chromen-2-one
Synonyms: limettin citropten
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: NXJCRELRQHZBQA-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C2C=CC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results