accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 487-21-8

IUPAC Name: 1,2,3,4-tetrahydropteridine-2,4-dione
Synonyms: pteridine-2,4-dione
Molecular Weight (g/mol): 164.12
Molecular Formula: C6H4N4O2
InChi Key: UYEUUXMDVNYCAM-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=NC=CN=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results