accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOHO

CAS RN 487-54-7

IUPAC Name: 2-[(2-hydroxyphenyl)formamido]acetic acid
Synonyms: salicyluric acid N-salicyloylglycine salicyloylglycine salicylurate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: ONJSZLXSECQROL-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C1=CC=CC=C1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results