accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 487-89-8

IUPAC Name: 1H-indole-3-carbaldehyde
Synonyms: indole-3-carboxaldehyde
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: OLNJUISKUQQNIM-UHFFFAOYSA-N
SMILES: O=CC1=CNC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (5)
  • (2)
Quantity 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (3)
  • (5)