accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4877-93-4

IUPAC Name: 2,2'-dimethoxy-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 2,2'-dimethoxy-
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: VGMKUVCDINAAFC-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CC=CC=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results