accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-47-1

IUPAC Name: tetrabromobenzene-1,2-diol
Synonyms: pyrocatechol, tetrabromo-
Molecular Weight (g/mol): 425.7
Molecular Formula: C6H2Br4O2
InChi Key: OAUWOBSDSJNJQP-UHFFFAOYSA-N
SMILES: OC1=C(O)C(Br)=C(Br)C(Br)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results