accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 488-48-2

IUPAC Name: tetrabromocyclohexa-2,5-diene-1,4-dione
Synonyms: bromanyl
Molecular Weight (g/mol): 423.68
Molecular Formula: C6Br4O2
InChi Key: LWHDQPLUIFIFFT-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(=O)C(Br)=C(Br)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results