accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOO

CAS RN 488-93-7

IUPAC Name: furan-3-carboxylic acid
Synonyms: 3-furoic acid
Molecular Weight (g/mol): 112.08
Molecular Formula: C5H4O3
InChi Key: IHCCAYCGZOLTEU-UHFFFAOYSA-N
SMILES: OC(=O)C1=COC=C1
Percent Purity 
  • (2)
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (2)