accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4884-24-6

IUPAC Name: [1,1'-bi(cyclopentane)]-2-one
Synonyms: bicyclopentylone
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: CWZGKTMWPFTJCS-UHFFFAOYNA-N
SMILES: O=C1CCCC1C1CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results