accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 4891-38-7

IUPAC Name: methyl 3-phenylprop-2-ynoate
Synonyms: methyl 3-phenylprop-2-ynoate
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: JFGWPXKGINUNDH-UHFFFAOYSA-N
SMILES: COC(=O)C#CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)