accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNH2NHO

CAS RN 4892-89-1

IUPAC Name: 1-[2-(morpholin-4-ium-4-yl)ethyl]piperazine-1,4-diium
Synonyms: 1-[2-(morpholin-4-ium-4-yl)ethyl]piperazine-1,4-diium
Molecular Weight (g/mol): 202.32
Molecular Formula: C10H24N3O
InChi Key: SAJZEJMFAWZNCQ-UHFFFAOYSA-Q
SMILES: C(C[NH+]1CCOCC1)[NH+]1CC[NH2+]CC1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)