accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 490-46-0

IUPAC Name: (2R,3R)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Synonyms: ent-epicatechin epi-catechin epi-catechol (-)-epicatechol l-acacatechin epicatechol α-catechin epicatechol, (-)- l-epicatechol L(-)-epicatechin
Molecular Weight (g/mol): 290.27
Molecular Formula: C15H14O6
InChi Key: PFTAWBLQPZVEMU-UKRRQHHQSA-N
SMILES: O[C@@H]1CC2=C(O)C=C(O)C=C2O[C@@H]1C1=CC=C(O)C(O)=C1
Grade 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)