accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 490-75-5

IUPAC Name: 3-(carboxymethyl)pyridine-2-carboxylic acid
Synonyms: homoquinolinic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: HQPMJFFEXJELOQ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CN=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results