CAS RN 490023-37-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sigma Aldrich (S)-1-{(Sp)-2-[2-(Diphenylphosphino)Phenyl]Ferrocenyl}Ethylbis[3,5-Bis-(Trifluoromethyl)Phenyl]Phosphine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.