accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClClCl

CAS RN 4901-51-3

IUPAC Name: 2,3,4,5-tetrachlorophenol
Synonyms: 2,3,4,5-tetrachlorophenol
Molecular Weight (g/mol): 231.88
Molecular Formula: C6H2Cl4O
InChi Key: RULKYXXCCZZKDZ-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results