accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4905-83-3

IUPAC Name: propan-1-amine hydrobromide
Synonyms: propylamine hydrobromide
Molecular Weight (g/mol): 140.02
Molecular Formula: C3H10BrN
InChi Key: MVYQJCPZZBFMLF-UHFFFAOYSA-N
SMILES: Br.CCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results