CAS RN 4906-68-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ AKR1C1 Inhibitor, 5-PBSA
The AKR1C1 Inhibitor, 5-PBSA, also referenced under CAS 4906-68-7, controls the biological activity of AKR1C1.