accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 491-38-3

IUPAC Name: 4H-chromen-4-one
Synonyms: 4H-1-benzopyran-4-one
Molecular Weight (g/mol): 146.15
Molecular Formula: C9H6O2
InChi Key: OTAFHZMPRISVEM-UHFFFAOYSA-N
SMILES: O=C1C=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results