accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 491-45-2

IUPAC Name: [1,1'-biphenyl]-2,3',4,5',6-pentol
Synonyms: [1,1'-biphenyl]-2,3',4,5',6-pentol
Molecular Weight (g/mol): 234.21
Molecular Formula: C12H10O5
InChi Key: KICYRZIVKKYRFS-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C(C(O)=C1)C1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results