accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3H3C(E)

CAS RN 4914-92-5

IUPAC Name: (2E)-3,4-dimethylpent-2-ene
Synonyms: 2-pentene, 3,4-dimethyl-, (E)-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: PPBWEVVDSRKEIK-FNORWQNLSA-N
SMILES: C\C=C(/C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results