accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 492-86-4

IUPAC Name: 2-(4-chlorophenyl)-2-hydroxyacetic acid
Synonyms: (4-chlorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: BWSFWXSSALIZAU-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)