accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 492-99-9

IUPAC Name: N-[(1E,2E)-2-(hydroxyimino)cyclohexylidene]hydroxylamine
Synonyms: N-[(1E,2E)-2-(hydroxyimino)cyclohexylidene]hydroxylamine
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: CUNNCKOPAWXYDX-KQQUZDAGSA-N
SMILES: O\N=C1/CCCC/C/1=N\O
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)