accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4920-84-7

IUPAC Name: 2,4-dimethoxy-1-nitrobenzene
Synonyms: 2,4-dimethoxy-1-nitro-benzene
Molecular Weight (g/mol): 183.16
Molecular Formula: C8H9NO4
InChi Key: XXWIYOBCHKCWNT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(OC)=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results