accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4920-95-0

IUPAC Name: 3,3',4,4'-tetramethyl-1,1'-biphenyl
Synonyms: 3,3',4,4'-tetramethyl-1,1'-biphenyl
Molecular Weight (g/mol): 210.32
Molecular Formula: C16H18
InChi Key: YXBIAYXZUDJVEB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)C1=CC=C(C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results