accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NBrCH3

CAS RN 4926-26-5

IUPAC Name: 2-bromo-4,6-dimethylpyridine
Synonyms: 2-bromo-4,6-dimethylpyridine
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: IRTOCXBLUOPRFT-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results