accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHNN

CAS RN 4928-87-4

IUPAC Name: 1H-1,2,4-triazole-5-carboxylic acid
Synonyms: 2H-1,2,4-triazole-3-carboxylic acid
Molecular Weight (g/mol): 113.08
Molecular Formula: C3H3N3O2
InChi Key: LJVQHXICFCZRJN-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results