CAS RN 493-72-1
IUPAC Name:
(5R)-3-hydroxy-5-phenylcyclohex-2-en-1-one
Synonyms:
(5R)-3-hydroxy-5-phenylcyclohex-2-en-1-one
Molecular Weight (g/mol):
188.23
Molecular Formula:
C12H12O2
InChi Key:
ZFVDSCASCRHXCP-SNVBAGLBSA-N
SMILES:
OC1=CC(=O)C[C@@H](C1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
5-Phenylcyclohexane-1,3-dione, 97%
5-Phenylcyclohexane-1,3-dione, 97%
CAS: 493-72-1 Molecular Formula: C12H12O2 Molecular Weight (g/mol): 188.23 MDL Number: MFCD00051846 InChI Key: ZFVDSCASCRHXCP-SNVBAGLBSA-N PubChem CID: 568962 IUPAC Name: 5-phenylcyclohexane-1,3-dione SMILES: OC1=CC(=O)C[C@@H](C1)C1=CC=CC=C1