accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNN

CAS RN 493-77-6

IUPAC Name: triphenyl-1,3,5-triazine
Synonyms: triphenyl-1,3,5-triazine
Molecular Weight (g/mol): 309.37
Molecular Formula: C21H15N3
InChi Key: HBQUOLGAXBYZGR-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)