accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4930-98-7

IUPAC Name: 2-hydrazinylpyridine
Synonyms: 2(1H)-pyridinone, hydrazone
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: NWELCUKYUCBVKK-UHFFFAOYSA-N
SMILES: NNC1=CC=CC=N1
Boiling Point 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)