accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 493024-39-8

IUPAC Name: 1-(bromomethyl)-3-chloro-5-fluorobenzene
Synonyms: 1-(bromomethyl)-3-chloro-5-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: PKSAHOPFPPAUOD-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC(CBr)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)