accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 494-90-6

IUPAC Name: (6R)-3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
Synonyms: menthofuran (R)-isomer menthofuran menthofuran, (R)- menthofuran, (r)-isomer menthofuran, (r)- tetrahydro-dimethylbenzofuran (r)-menthofuran menthofuran, (R)-isomer (+)-menthofuran (r)-isomer menthofuran
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: YGWKXXYGDYYFJU-SSDOTTSWSA-N
SMILES: C[C@@H]1CCC2=C(C1)OC=C2C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)