accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3S

CAS RN 4946-14-9

IUPAC Name: [4-(propan-2-yl)phenyl]sulfanide
Synonyms: (4-isopropylphenyl)sulfanide
Molecular Weight (g/mol): 151.25
Molecular Formula: C9H11S
InChi Key: APDUDRFJNCIWAG-UHFFFAOYSA-M
SMILES: CC(C)C1=CC=C([S-])C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results