accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 495-07-8

IUPAC Name: 2-(chloromethyl)-1,4-difluorobenzene
Synonyms: 2-(chloromethyl)-1,4-difluorobenzene
Molecular Weight (g/mol): 162.56
Molecular Formula: C7H5ClF2
InChi Key: INXKTZMJFPRVAY-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results