accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 495-48-7

IUPAC Name: 1-oxo-1,2-diphenylhydrazin-1-ium-2-ide
Synonyms: 1-oxo-1,2-diphenylhydrazin-1-ium-2-ide
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: GAUZCKBSTZFWCT-UHFFFAOYSA-N
SMILES: O=[N+]([N-]C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)