accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHO

CAS RN 495-69-2

IUPAC Name: 2-(phenylformamido)acetic acid
Synonyms: hippuric acid (14)C-labeled hippuric acid monopotassium salt hippuric acid monoammonium salt hippuric acid hippuric acid, (14)c-labeled hippuric acid (substance) benzamidoessigsaeure hippursaeure benzoylaminoessigsaeure hippuric acid, (14)C-labeled
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: QIAFMBKCNZACKA-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (7)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (7)
  • (1)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)
  • (1)