accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSHN

CAS RN 49559-83-3

IUPAC Name: 5-methoxy-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 5-methoxy-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 181.21
Molecular Formula: C8H7NO2S
InChi Key: QGTUVLRFJOUWBN-UHFFFAOYSA-N
SMILES: COC1=CC=C2OC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results