accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHClHCl

CAS RN 49563-87-3

IUPAC Name: 1,4-diazabicyclo[2.2.2]octane dihydrochloride
Synonyms: dabco dihydrochloride
Molecular Weight (g/mol): 185.09
Molecular Formula: C6H14Cl2N2
InChi Key: FXNCMDQKBKIDGU-UHFFFAOYSA-N
SMILES: Cl.Cl.C1CN2CCN1CC2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results