CAS RN 4959-17-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ 7-Aminoclonazepam Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
This certified Snap-N-Spike™ solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology and urine drug testing.