accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHH3C

CAS RN 496-78-6

IUPAC Name: 2,4,5-trimethylphenol
Synonyms: 2,4,5-trimethylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: VXSCPERJHPWROZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results