accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4961-40-4

HHClHClHClClH2NHNNHNH2

CAS RN 4961-40-4

IUPAC Name: tetrahydrogen (2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine tetrachloride
Synonyms: tetrahydrogen trien tetrachloride
Molecular Weight (g/mol): 292.07
Molecular Formula: C6H22Cl4N4
InChi Key: OKHMDSCYUWAQPT-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NCCNCCNCCN
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)